2-[[2-(Dimethylamino)ethyl]methylamino]ethanol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
N'-(2-Hydroxyethyl)-N,N,N'-trimethylethylenediamine, N2-(2-Hydroxyethyl)-N1,N1,N2-trimethyl-1,2-ethylenediamine
D4680-100ML
D4680-25ML
TCD4680-100MLEA
107
USD
TCD4680-100ML
TCD4680-25ML
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol ≥97.0% (by GC, titration analysis)
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol
Formula:
C₇H₁₈N₂O MW: 146.23 g/mol Boiling Pt: 88°C Flash Pt: 87°C |
MDL Number:
MFCD00010387 CAS Number: 2212-32-0 EINECS: 218-658-4 |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...