2-(2-Bromophenyl)naphthalene ≥98.0% (by GC)
Supplier: TCI America
B6100-1G
B6100-5G
TCB6100-1GEA
149
USD
TCB6100-1G
TCB6100-5G
2-(2-Bromophenyl)naphthalene ≥98.0% (by GC)
2-(2-Bromophenyl)naphthalene
Formula:
C₁₆H₁₁Br |
MDL Number:
MFCD18415513 CAS Number: 22082-97-9 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...