9-(1-Naphthyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 9-(Naphthalen-1-yl)-9H-carbazole

N1012-1G N1012-5G
TCN1012-1GEA 159 USD
TCN1012-1G TCN1012-5G
9-(1-Naphthyl)carbazole ≥98.0% (by GC)
9-(1-Naphthyl)carbazole
Formula: C₂₂H₁₅N
MW: 293.37 g/mol
MDL Number: MFCD26127424
CAS Number: 22034-43-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR