5-Aminovaleric acid hydrobromide, low water content ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A3094-1G
A3094-5G
TCA3094-1GEA
111
USD
TCA3094-1G
TCA3094-5G
5-Aminovaleric acid hydrobromide, low water content ≥98.0% (by titrimetric analysis)
5-Aminovaleric acid hydrobromide
Formula:
C₅H₁₁NO₂·HBr |
CAS Number:
2173111-73-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Water | max. 250 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...