4-Aminocrotonic acid hydrochloride 97,0 HPLC_ASSAY_METHOD
Supplier: TCI America
|
Warning
|
Synonyms:
trans-4-Amino-2-butenoic acid hydrochloride
A3011-100MG
A3011-10MG
TCA3011-100MGEA
700
USD
TCA3011-100MG
TCA3011-10MG
4-Aminocrotonic acid hydrochloride 97,0 HPLC_ASSAY_METHOD
4-Aminocrotonic acid hydrochloride
Formula:
C₄H₇NO₂·HCl MW: 137.56 g/mol |
CAS Number:
2126899-84-9 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...