9-(4'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 9-(4'-Bromo-[1,1'-biphenyl]-4-yl)-9H-carbazole

B5109-1G B5109-5G
TCB5109-1GEA 37 USD
TCB5109-1G TCB5109-5G
9-(4'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)
9-(4'-Bromo-4-biphenylyl)carbazole
Formula: C₂₄H₁₆BrN
MW: 398.30 g/mol
MDL Number: MFCD27644692
CAS Number: 212385-73-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR