2,5-Di(2-thienyl)thieno[3,2-b]thiophene ≥94.0% (by GC)

Supplier: TCI America

Synonyms: 2,5-Di(thiophen-2-yl)thieno[3,2-b]thiophene

D4725-1G D4725-200MG
TCD4725-1GEA 535.6 USD
TCD4725-1G TCD4725-200MG
2,5-Di(2-thienyl)thieno[3,2-b]thiophene ≥94.0% (by GC)
2,5-Di(2-thienyl)thieno[3,2-b]thiophene
Formula: C₁₄H₈S₄
MW: 304.46 g/mol
MDL Number: MFCD11114556
CAS Number: 21210-90-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR