(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2907-1G M2907-5G
76220-292EA 116 USD
76220-292 76220-294
(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate
Formula: C₁₇H₁₈F₃IO₃S
MDL Number: MFCD20264884
CAS Number: 210823-54-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR