N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine ≥98.0% (by HPLC)

Supplier: TCI America

B5117-1G B5117-200MG
TCB5117-1GEA 248 USD
TCB5117-1G TCB5117-200MG
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine
Formula: C₆₀H₄₆N₄
CAS Number: 209980-53-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 248.0 to 252.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR