2-Methylpyridine-5-boronic acid hydrochloride (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
6-Methyl-3-pyridylboronic acid hydrochloride
M2253-1G
M2253-5G
TCM2253-1GEA
108.8
USD
TCM2253-1G
TCM2253-5G
2-Methylpyridine-5-boronic acid hydrochloride (contains varying amounts of Anhydride)
2-Methylpyridine-5-boronic acid hydrochloride
Formula:
C₆H₈BNO₂·HCl MW: 173.40 g/mol |
CAS Number:
2096333-73-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | 95.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...