1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)

Supplier: TCI America

B5364-1G B5364-5G
76219-594EA 88 USD
76219-594 76219-596
1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)
1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene
Formula: C₁₉H₂₈F₂O
MDL Number: MFCD20489249
CAS Number: 208709-55-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR