5-Bromo-1-methylisatin ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-1-methylindoline-2,3-dione

B4779-1G B4779-5G
TCB4779-1GEA 135 USD
TCB4779-1G TCB4779-5G
5-Bromo-1-methylisatin ≥98.0% (by GC)
5-Bromo-1-methylisatin
Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
MDL Number: MFCD00456313
CAS Number: 2058-72-2

Order Now

Specification Test Results

Appearance Light red to Yellow to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR