1-Ethyl-4-octylbenzene ≥97.0% (by GC)
Supplier: TCI America
E1431-5G
E1431-1G
TCE1431-5GEA
451
USD
TCE1431-5G
TCE1431-1G
1-Ethyl-4-octylbenzene ≥97.0% (by GC)
1-Ethyl-4-octylbenzene
Formula:
C₁₆H₂₆ |
CAS Number:
204008-22-0 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Specific gravity (20/20) | 0.8530 to 0.8630 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...