2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F1045-1G
F1045-200MG
76220-078EA
164
USD
76220-078
76220-080
2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)
2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine
Formula:
C₂₁H₁₄FN₃ MW: 327.36 g/mol |
MDL Number:
MFCD28964710 CAS Number: 203450-08-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 231.0 to 235.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...