1-Bromo-2,4,5-trimethoxybenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4721-1G B4721-200MG
76219-422EA 331 USD
76219-422 76219-424
1-Bromo-2,4,5-trimethoxybenzene ≥98.0% (by GC)
1-Bromo-2,4,5-trimethoxybenzene
Formula: C₉H₁₁BrO₃
Boiling Pt: 263°C
MDL Number: MFCD00094684
CAS Number: 20129-11-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR