Triethoxy(pentafluorophenyl)silane ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
(Triethoxysilyl)pentafluorobenzene, Triethoxy(perfluorophenyl)silane
T3134-1G
T3134-5G
TCT3134-1GEA
60
USD
TCT3134-1G
TCT3134-5G
Triethoxy(pentafluorophenyl)silane ≥95.0% (by GC)
Triethoxy(pentafluorophenyl)silane
Formula:
C₁₂H₁₅F₅O₃Si MW: 330.33 g/mol Boiling Pt: 238°C Flash Pt: 104°C Storage Temperature: Refrigerator |
MDL Number:
MFCD03411257 CAS Number: 20083-34-5 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...