(2-Hydroxy-5-methylphenyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
H1749-5G
H1749-1G
TCH1749-5GEA
218
USD
TCH1749-5G
TCH1749-1G
(2-Hydroxy-5-methylphenyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
(2-Hydroxy-5-methylphenyl)triphenylphosphonium iodide
Formula:
C₂₅H₂₂IOP |
CAS Number:
2005487-70-5 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Appearance | White to Almost white powder to crystal |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...