(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
MeTAPS-Br
H1748-1G
H1748-5G
TCH1748-1GEA
66
USD
TCH1748-1G
TCH1748-5G
(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide
Formula:
C₂₅H₂₂BrOP MW: 449.33 g/mol |
CAS Number:
2005487-65-8 |
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...