4'-Butyl-4-iodobiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Butyl-4'-iodobiphenyl

B5211-1G B5211-200MG
TCB5211-1GEA 220 USD
TCB5211-1G TCB5211-200MG
4'-Butyl-4-iodobiphenyl ≥98.0% (by GC)
4'-Butyl-4-iodobiphenyl
Formula: C₁₆H₁₇I
MW: 336.22 g/mol
MDL Number: MFCD16619281
CAS Number: 199982-02-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 122.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR