3,3'-Bicarbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
Synonyms:
9H,9'H-3,3'-Bicarbazole
B4646-1G
B4646-200MG
TCB4646-1GEA
191
USD
TCB4646-1G
TCB4646-200MG
3,3'-Bicarbazole ≥98.0% (by HPLC, total nitrogen)
3,3'-Bicarbazole
Formula:
C₂₄H₁₆N₂ MW: 332.41 g/mol |
MDL Number:
MFCD11046470 CAS Number: 1984-49-2 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...