2-Hexyl-1-n-octanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Hexyl-n-octyl alcohol

H1635-1ML H1635-5ML
TCH1635-1MLEA 148 USD
TCH1635-1ML TCH1635-5ML
2-Hexyl-1-n-octanol ≥98.0% (by GC)
2-Hexyl-1-n-octanol
Formula: C₁₄H₃₀O
MW: 214.39 g/mol
Boiling Pt: 162°C
MDL Number: MFCD00046769
CAS Number: 19780-79-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR