1,3-Dibromo-5-iodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4691-1G D4691-5G
TCD4691-1GEA 43 USD
TCD4691-1G TCD4691-5G
1,3-Dibromo-5-iodobenzene ≥98.0% (by GC)
1,3-Dibromo-5-iodobenzene
Formula: C₆H₃Br₂I
MDL Number: MFCD07778996
CAS Number: 19752-57-9

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR