1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Cyclohexylcarbamic acid 1,2-Bis(4-methoxyphenyl)-2-oxoethyl ester, 1,2-Bis(4-methoxyphenyl)-2-oxoethyl cyclohexylcarbamate

B5085-1G B5085-200MG
TCB5085-1GEA 354.6 USD
TCB5085-1G TCB5085-200MG
1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate ≥98.0% (by HPLC, total nitrogen)
1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate
Formula: C₂₃H₂₇NO₅
MW: 397.47 g/mol
MDL Number: MFCD29472516
CAS Number: 196599-80-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR