1,3-Benzenediboronic acid bis(pinacol) ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,3-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

B5070-1G B5070-5G
76218-806EA 118 USD
76218-806 76218-808
1,3-Benzenediboronic acid bis(pinacol) ester ≥98.0% (by GC)
1,3-Benzenediboronic acid bis(pinacol) ester
Formula: C₁₈H₂₈B₂O₄
MDL Number: MFCD09864185
CAS Number: 196212-27-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 133.0 to 137.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR