N,N,N'-Triphenyl-1,4-phenylenediamine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3367-1G T3367-200MG
TCT3367-1GEA 175 USD
TCT3367-1G TCT3367-200MG
N,N,N'-Triphenyl-1,4-phenylenediamine ≥98.0% (by GC)
N,N,N'-Triphenyl-1,4-phenylenediamine
Formula: C₂₄H₂₀N₂
MDL Number: MFCD27939629
CAS Number: 19606-98-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR