Dipyrido[3,2-a:2',3'-c]phenazine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Dppz

D4379-1G D4379-200MG
TCD4379-1GEA 253 USD
TCD4379-1G TCD4379-200MG
Dipyrido[3,2-a:2',3'-c]phenazine ≥98.0% (by HPLC)
Dipyrido[3,2-a:2',3'-c]phenazine
Formula: C₁₈H₁₀N₄
MW: 282.31 g/mol
Melting Pt: 248°C
MDL Number: MFCD04113602
CAS Number: 19535-47-8

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR