4-Bromo-4',4''-dimethoxytriphenylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B4667-1G B4667-5G
TCB4667-1GEA 145 USD
TCB4667-1G TCB4667-5G
4-Bromo-4',4''-dimethoxytriphenylamine ≥98.0% (by HPLC, total nitrogen)
4-Bromo-4',4''-dimethoxytriphenylamine
Formula: C₂₀H₁₈BrNO₂
MDL Number: MFCD22124385
CAS Number: 194416-45-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR