2-(4-Bromophenyl)benzo[b]thiophene ≥98.0% (by GC)

Supplier: TCI America

B5016-1G B5016-200MG
76219-484EA 251 USD
76219-484 76219-486
2-(4-Bromophenyl)benzo[b]thiophene ≥98.0% (by GC)
2-(4-Bromophenyl)benzo[b]thiophene
Formula: C₁₄H₉BrS
MW: 289.19 g/mol
MDL Number: MFCD22380322
CAS Number: 19437-86-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 213.0 to 217.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR