Bis(4-pyridyl)amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,4'-Dipyridylamine, N-(4-Pyridinyl)-4-pyridinamine, Di(pyridin-4-yl)amine

B6124-1G B6124-250MG
TCB6124-1GEA 249 USD
TCB6124-1G TCB6124-250MG
Bis(4-pyridyl)amine ≥98.0% (by GC, titration analysis)
Bis(4-pyridyl)amine
Formula: C₁₀H₉N₃
MW: 171.20 g/mol
MDL Number: MFCD00829434
CAS Number: 1915-42-0

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR