Calcium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C3476-1G C3476-5G
TCC3476-1GEA 166 USD
TCC3476-1G TCC3476-5G
Calcium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)
Calcium bis(fluorosulfonyl)imide
Formula: CaF₄N₂O₈S₄
CAS Number: 1906900-37-5
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR