N,N-Dipropargyl-p-toluenesulfonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
4-Methyl-N,N-di(prop-2-yn-1-yl)benzenesulphonamide
D4964-1G
D4964-5G
TCD4964-1GEA
137
USD
TCD4964-1G
TCD4964-5G
N,N-Dipropargyl-p-toluenesulfonamide ≥98.0% (by HPLC, total nitrogen)
N,N-Dipropargyl-p-toluenesulfonamide
Formula:
C₁₃H₁₃NO₂S MW: 247.31 g/mol |
MDL Number:
MFCD22575206 CAS Number: 18773-54-1 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...