Bis(4-bromophenyl)diphenylsilane ≥98.0% (by GC)

Supplier: TCI America

B5087-1G B5087-200MG
TCB5087-1GEA 501 USD
TCB5087-1G TCB5087-200MG
Bis(4-bromophenyl)diphenylsilane ≥98.0% (by GC)
Bis(4-bromophenyl)diphenylsilane
Formula: C₂₄H₁₈Br₂Si
MDL Number: MFCD29089347
CAS Number: 18733-91-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR