Bis(triethoxysilyl)methane ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
1,1,1,3,3,3-Hexaethoxy-1,3-disilapropane, 4,4,6,6-Tetraethoxy-3,7-dioxa-4,6-disilanonane
T3936-1G
T3936-5G
TCT3936-1GEA
61
USD
TCT3936-1G
TCT3936-5G
Bis(triethoxysilyl)methane ≥95.0% (by GC)
Bis(triethoxysilyl)methane
Formula:
C₁₃H₃₂O₆Si₂ MW: 340.56 g/mol Boiling Pt: 114°C |
MDL Number:
MFCD00270971 CAS Number: 18418-72-9 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...