2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 2,4-Di([1,1'-biphenyl]-4-yl)-6-chloro-1,3,5-triazine

B4941-1G B4941-200MG
76219-458EA 380 USD
76219-458 76219-460
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine
Formula: C₂₇H₁₈ClN₃
MW: 419.91 g/mol
MDL Number: MFCD27979765
CAS Number: 182918-13-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR