Isopropoxytrimethylsilane ≥98.0% (by GC)

Supplier: TCI America
Danger

I1133-25ML I1133-5ML
TCI1133-25MLEA 104 USD
TCI1133-25ML TCI1133-5ML
Isopropoxytrimethylsilane ≥98.0% (by GC)
Isopropoxytrimethylsilane
Formula: C₆H₁₆OSi
Boiling Pt: 87°C
Flash Pt: -11°C
MDL Number: MFCD00134520
CAS Number: 1825-64-5
EINECS: 217-372-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR