1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

T3356-1G T3356-5G
TCT3356-1GEA 59 USD
TCT3356-1G TCT3356-5G
1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane ≥97.0% (by HPLC, total nitrogen)
1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane
Formula: C₃₆H₄₄N₄
MW: 532.78 g/mol
MDL Number: MFCD05864401
CAS Number: 18084-64-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 143.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR