Tri(cyclohexa-2,5-dien-1-yl)silane ≥93.0% (by GC)

Supplier: TCI America
Warning

T3443-1G T3443-200MG
TCT3443-1GEA 204 USD
TCT3443-1G TCT3443-200MG
Tri(cyclohexa-2,5-dien-1-yl)silane ≥93.0% (by GC)
Tri(cyclohexa-2,5-dien-1-yl)silane
Formula: C₁₈H₂₂Si
MW: 266.46 g/mol
CAS Number: 1807776-35-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 93.0 %
Melting point 63.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR