Yoshimulactone Green (YLG) ≥98.0% (by HPLC)

Supplier: TCI America

E1238-10MG E1238-1MG
TCE1238-10MGEA 80 USD
TCE1238-10MG TCE1238-1MG
Yoshimulactone Green (YLG) ≥98.0% (by HPLC)
Yoshimulactone Green (YLG)
Formula: C₂₇H₂₀O₇
CAS Number: 1807549-00-3

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 188.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR