1-(Trimethoxysilyl)naphthalene ≥98.0% (by GC)

Supplier: TCI America
Warning

T3292-1G T3292-5G
TCT3292-1GEA 48.1 USD
TCT3292-1G TCT3292-5G
1-(Trimethoxysilyl)naphthalene ≥98.0% (by GC)
1-(Trimethoxysilyl)naphthalene
Formula: C₁₃H₁₆O₃Si
Boiling Pt: 148°C
Storage Temperature: Refrigerator
MDL Number: MFCD00094090
CAS Number: 18052-76-1

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR