Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Ethyl(dimethyl)phenethylammonium bis(trifluoromethanesulphonyl)imide

E1283-1G E1283-5G
TCE1283-1GEA 105 USD
TCE1283-1G TCE1283-5G
Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulfonyl)imide
Formula: C₁₄H₂₀F₆N₂O₄S₂
MW: 458.43 g/mol
CAS Number: 1804970-28-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (HPLC) min. 96.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR