1,3-Diallylurea ≥98.0% (by GC)

Supplier: TCI America

D5170-1G D5170-5G
76219-214EA 162 USD
76219-214 76219-216
1,3-Diallylurea ≥98.0% (by GC)
1,3-Diallylurea
Formula: C₇H₁₂N₂O
Boiling Pt: 56°C
MDL Number: MFCD00008639
CAS Number: 1801-72-5
EINECS: 217-291-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR