5,6-Diamino-1,3-diethyluracil hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5,6-Diamino-1,3-diethylpyrimidine-2,4(1H,3H)-dione hydrochloride
D4659-1G
D4659-5G
TCD4659-1GEA
104.6
USD
TCD4659-1G
TCD4659-5G
5,6-Diamino-1,3-diethyluracil hydrochloride ≥98.0% (by HPLC, titration analysis)
5,6-Diamino-1,3-diethyluracil hydrochloride
Formula:
C₈H₁₄N₄O₂·HCl MW: 234.68 g/mol |
MDL Number:
MFCD22573561 CAS Number: 1785764-26-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 235.0 to 239.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...