4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Parylene F Dimer, 5,6,11,12,13,14,15,16-Octafluorotricyclo[8.2.2.2(4,7)]hexadeca-4,6,10,12,13,15-hexaene

O0455-1G O0455-5G
TCO0455-1GEA 82.6 USD
TCO0455-1G TCO0455-5G
4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane ≥98.0% (by GC)
4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane
Formula: C₁₆H₈F₈
MW: 352.23 g/mol
MDL Number: MFCD13195584
CAS Number: 1785-64-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR