9,10-Dibromo-2-methylanthracene ≥96.0% (by GC)

Supplier: TCI America

D4977-1G D4977-200MG
TCD4977-1GEA 446 USD
TCD4977-1G TCD4977-200MG
9,10-Dibromo-2-methylanthracene ≥96.0% (by GC)
9,10-Dibromo-2-methylanthracene
Formula: C₁₅H₁₀Br₂
MDL Number: MFCD00266492
CAS Number: 177839-45-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR