9,10-Bis[N,N-di(p-tolyl)amino]anthracene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N9,N9,N10,N10-Tetra-p-tolylanthracene-9,10-diamine, N,N,N',N'-Tetrakis(4-methylphenyl)-9,10-anthracenediamine

B4966-1G B4966-200MG
TCB4966-1GEA 429 USD
TCB4966-1G TCB4966-200MG
9,10-Bis[N,N-di(p-tolyl)amino]anthracene ≥98.0% (by HPLC)
9,10-Bis[N,N-di(p-tolyl)amino]anthracene
Formula: C₄₂H₃₆N₂
MW: 568.76 g/mol
MDL Number: MFCD12022465
CAS Number: 177799-16-5

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR