2-Iododibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

I0973-1G I0973-200MG
TCI0973-1GEA 251 USD
TCI0973-1G TCI0973-200MG
2-Iododibenzothiophene ≥98.0% (by GC)
2-Iododibenzothiophene
Formula: C₁₂H₇IS
MW: 310.15 g/mol
Melting Pt: 88°C
CAS Number: 177586-41-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 89.0 to 93.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR