Ambrisentan ≥98.0% (by HPLC)

Supplier: TCI America
Warning

A2966-100MG A2966-20MG
76218-768EA 84 USD
76218-768 76218-770
Ambrisentan ≥98.0% (by HPLC)
Ambrisentan
Formula: C₂₂H₂₂N₂O₄
MDL Number: MFCD09842330
CAS Number: 177036-94-1
Merck Index: 14,00384

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.00 to 7.80 %
Specific rotation [a]20/D +170 to +176 deg (C=0.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR