Dimethyl 2,2'-Bipyridine-5,5'-dicarboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

D5024-1G D5024-5G
TCD5024-1GEA 97 USD
TCD5024-1G TCD5024-5G
Dimethyl 2,2'-Bipyridine-5,5'-dicarboxylate ≥98.0% (by GC)
Dimethyl 2,2'-Bipyridine-5,5'-dicarboxylate
Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD30531632
CAS Number: 1762-45-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR