1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)

Supplier: TCI America

E1230-1G E1230-5G
76220-038EA 52 USD
76220-038 76220-040
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene
Formula: C₁₇H₂₄F₂O
MDL Number: MFCD11053400
CAS Number: 174350-05-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 57.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR