Potassium Phenyl sulfate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P0232-1G P0232-5G
TCP0232-1GEA 155 USD
TCP0232-1G TCP0232-5G
Potassium Phenyl sulfate ≥98.0% (by HPLC, titration analysis)
Potassium Phenyl sulfate
Formula: C₆H₅KO₄S
Melting Pt: 160°C
Storage Temperature: Freezer
CAS Number: 1733-88-6
EINECS: 217-071-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR